Order Entry
Finland
ContactUsLinkComponent
Anti-CD8A Mouse Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: OKT8]
Anti-CD8A Mouse Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: OKT8]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD8A Mouse Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: OKT8]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 65-0086-T025
  • 65-0086-T100
  • Antigen symbol:
  • CD8A
  • CD8A
  • Antigen name:
  • CD8 antigen, alpha chain
  • CD8 antigen, alpha chain
  • Antigen synonyms:
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • p32|Lyt-66|BB154331|Ly-2|Leu2|Ly-35|MAL|CD8|Ly-B
  • Conjugation:
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
  • Clone:
  • OKT8
  • OKT8
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 925
  • 925
  • Isotype:
  • IgG2a
  • IgG2a
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Store Undiluted at 2...8 °C; Protect from Light; Do Not Freeze.
  • Store Undiluted at 2...8 °C; Protect from Light; Do Not Freeze.
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications