Battery charger kit TR-644E, Versaflo™
Catalog # MMMATR-644E
Supplier: 3M
Specifications
- Description:Battery charger kit TR-644E
- Pk:1
Specifications
About this item
The Versaflo™ Battery Charger Kit TR-644E is a four-station battery charger designed for use with all batteries from the Versaflo™ Powered Air Turbo TR-600 Series. It includes four TR-640 charging cradles, a baseplate, power supply, and lead.
- Charges in under four hours
- Charges up to four batteries at a time
- Store fully-charged batteries on charger, ready for immediate use
- CE approved
This battery kit will charge four standard batteries (TR-630), or high-capacity batteries (TR-632) in less than 4 hours, which can then be stored on the cradle once fully charged, ready for immediate use. The kit includes four charging cradles, a baseplate, power supply, and lead.