About this item
GenScript's Bis-Tris gels are high-performing polyacrylamide gels designed to separate a wide range of protein sizes by electrophoresis. The gels are cast in a neutral pH buffer that minimises polyacrylamide hydrolysis and increases gel stability. Furthermore, the neutral pH which minimises protein modification when compared to Tris-glycine gels.
- Large well volume
- High resoluton and reproducibility
- Long shelf life
- Cost effective
- Compatible cassette design
GenScript's SurePAGE™ Gels are premium, high resolution with enhance reproducibility gels, while the ExpressPlus are good quality and cost-effective gels.
Proteins are separated on SurePAGE using MOPS or MES running buffer. We recommend using MOPS buffer for protein sizes that are >30 kD and MES for proteins that are <30 kD.
We recommend using 4×LDS sample buffer (M00676) as the sample loading buffer.