Order Entry
Finland
ContactUsLinkComponent
Hand-Operated Drum Pumps, Steel
Hand-Operated Drum Pumps, Steel
Certificates
undefined
Hand-Operated Drum Pumps, Steel
Specifications
  • Tube ext.Ø, inlet:
  • 0,625"
  • 0,625"
  • 0,625"
  • 0,625"
  • 0,625"
  • 0,625"
  • Tube ext.Ø, outlet:
  • 0,5"
  • 0,5"
  • 0,5"
  • 0,5"
  • 1"
  • 1"
  • Tube length, inlet:
  • 23"
  • 23"
  • 37"
  • 37"
  • Wetted materials:
  • 316 SS body; PTFE packing, piston, inlet tube
  • Carbon steel body; HDPE packing, piston, inlet tube
  • 317 SS body; PTFE packing, piston, inlet tube
  • Carbon steel body; HDPE packing, piston, inlet tube
  • Steel, polyethylene
  • 304 SS, PTFE
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
  • 1 year
  • 3 Months
  • 3 Months
Specifications