Order Entry
Finland
ContactUsLinkComponent
Heavy-Duty Tubing Pinch Clamps, Polysulfone
Heavy-Duty Tubing Pinch Clamp, Polysulfone
Catalog # MFLX20001-00
undefined
Heavy-Duty Tubing Pinch Clamp, Polysulfone
Catalog # MFLX20001-00
Specifications
  • Description:
    Heavy-Duty Tubing Pinch Clamp, Polysulfone
  • Material:
    Polysulfone
  • Warranty:
    3 Months
  • Pk:
    1
Specifications
About this item
Durable design handles higher working pressures.
  • Quick installation with positive swing-away locking screw
  • Smooth contours allow for easy cleaning
Use this clamp with tubing or hose - it will meet the working pressure of either. High-strength polysulfone closure provides excellent durability, can be continually reused, and resists most chemicals. Simply align the hose within the clamp; mating surfaces on the clamp indicate complete closure. Typical applications include sterile filling, media bag connections, pumping, liquid control and high-purity fluid transfers. FDA-compliant polysulfone material can be sterilized by autoclave or gamma radiation.
  • Description:
    Heavy-Duty Tubing Pinch Clamp, Polysulfone
  • Material:
    Polysulfone
  • Environmentally Preferable:
  • Warranty:
    3 Months
  • Pk:
    1