Pre-sterilized individually wrapped blue pestle, Axygen®
Catalog # AXYGPES-15-B-SI
Supplier: Corning
Specifications
- For:1.5 - 2.0 ml
- Description:Pre-sterilized, individually wrapped pestle
- Pk:400
Specifications
About this item
Durable and inert polypropylene construction
- Designed to fit 1.5 and 2.0 ml centrifuge tubes
- Individually wrapped and sterilized