Order Entry
Finland
ContactUsLinkComponent
 
Terrific broth, Corning®
Catalog # MDTC46-055-CM
Supplier:  Corning
CAS Number:  
undefined
Terrific broth, Corning®
Catalog # MDTC46-055-CM
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Description:
    Terrific broth12.00 g casein peptone, 4.00 (ml) glyerol, 2.31 g KH₂PO₄, 12.54 g KH₂PO₄, 24.00 g yeastolate, 1 L
  • Storage temperature:
    Ambient
  • Shipping temperature:
    Ambient

Specifications

About this item

Terrific broth is a medium for the propagation and maintenance of recombinant E.coli.

Because this medium supports higher cell densities than LB medium, it is preferred for maximizing plasmid yield in preparation for nucleic acid purification.