Order Entry
Finland
ContactUsLinkComponent
Pressure Relief Valves, 316 Stainless Steel
Pressure Relief Valve, 316 SS, 1/2" NPT(F)
Catalog # MFLX03245-40
undefined
Pressure Relief Valve, 316 SS, 1/2" NPT(F)
Catalog # MFLX03245-40
Specifications
  • Connection Type:
    NPT(F)
  • Max. pressure:
    150 psi
  • Description:
    Pressure Relief Valve, 316 SS, 1/2" NPT(F)
  • Wetted materials:
    Stainless Steel
  • Warranty:
    1 year
  • Orifice:
    7,9375 mm (0,3125")
  • Pk:
    1
Specifications
About this item
Easily control your outlet pressure.
  • Constructed of durable 316 stainless steel (SS)
Use for liquid applications with flow rates up to 15 GPM. For pressures from 40 to 150 psig. Valves have 1/2" NPT(F); orifice size is 5/16".
  • Connection Type:
    NPT(F)
  • Max. pressure:
    150 psi
  • Description:
    Pressure Relief Valve, 316 SS, 1/2" NPT(F)
  • Environmentally Preferable:
  • Wetted materials:
    Stainless Steel
  • Warranty:
    1 year
  • Orifice:
    7,9375 mm (0,3125")
  • Pk:
    1