Order Entry
Finland
ContactUsLinkComponent
Wide-Range Pressure Relief Valves
Wide-Range Pressure Relief Valves
Certificates
undefined
Wide-Range Pressure Relief Valves
Specifications
  • Max. pressure:
  • 210 psi
  • 14,5 bar (210 psi)
  • 14,5 bar (210 psi)
  • Media compatibility:
  • Liquids
  • Liquids
  • Liquids
  • Wetted materials:
  • PVC and Viton
  • PVC and Viton
  • PTFE and Viton
  • Warranty:
  • 1 year
  • 1 year
  • 1 year
Specifications