Order Entry
Finland
ContactUsLinkComponent
Scienceware Flow Control Hosecock Clamps, Plastic
Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
Catalog # MFLX06833-10
undefined
Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
Catalog # MFLX06833-10
Specifications
  • Description:
    Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
  • Clamp material:
    Delrin acetal
  • Pk:
    3
  • Clamp type:
    Tubing
  • Max. grip range:
    0,5"
Specifications
About this item
Deliver accurate regulation or interruption of fluid flow.
  • Constructed of Delrin acetal
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.
  • Description:
    Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
  • Environmentally Preferable:
  • Clamp material:
    Delrin acetal
  • Pk:
    3
  • Clamp type:
    Tubing
  • Max. grip range:
    0,5"