Order Entry
Finland
ContactUsLinkComponent
Neomycin sulphate, powder
Neomycin sulphate, powder
Catalog # MDTC61-241-RG
Supplier:  Corning
CAS Number:  
undefined
Neomycin sulphate, powder
Catalog # MDTC61-241-RG
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :908.87 g/mol
Storage Temperature:Ambient
CAS Number:1405-10-3
 :MFCD00077833
Formula:C₂₃H₄₆N₆O₁₃·3H₂SO₄
MDL Number:MFCD00150827