Order Entry
Finland
ContactUsLinkComponent
Kanamycin sulphate, powder
Kanamycin sulphate, powder
Catalog # MDTC61-176-RG
Supplier:  Corning
CAS Number:  
undefined
Kanamycin sulphate, powder
Catalog # MDTC61-176-RG
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :582.6 g/mol
Storage Temperature:Refrigerator
CAS Number:25389-94-0
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253