Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Penicillin/Streptomycin/Glutamine
Penicillin/Streptomycin/Glutamine 100X, Corning®
Penicillin/Streptomycin/Glutamine 100X, Corning®
Penicillin/Streptomycin/Glutamine 100X, Corning®
Catalog # MDTC30-009-CI
Supplier:
Corning
CAS Number:
Certificates
Expand image
Penicillin/Streptomycin/Glutamine 100X, Corning®
Catalog # MDTC30-009-CI
Supplier:
Corning
CAS Number:
Certificates
Specifications
Pack type:
Plastic bottle
Pk:
6
Seq:
2455
Pk Info:
6 x 100 mL
Specifications
About this item
Product Details
Documentation
:
Storage Temperature:
Freezer
Product Details
Documentation