Ultra fine smear-proof markers
Supplier: MICRONOVA
Certificates
About this item
A permanent marker that gives the thinnest permanent line at 0,3 mm.
- Long nib allows for writing in limited spaces
- Works well with lab samples and slides, glass, plastic, and paper
- Gamma irradiated for aseptic environments
- Available in black or blue ink
Writes permanently on most surfaces with quick drying ink that is fade- and water-resistant to further extend multiple project uses. Designed for documentation and labeling in aseptic environments.