Conical tubes, bulk, sterile, Thermo Scientific™
Supplier: Thermo Fisher Scientific
Certificates
About this item
The Thermo Scientific™ Conical Centrifuge Tubes are sterile and ideal for applications requiring larger sample volumes.
- Manufactured with high quality virgin resins
- Sterilised by gamma irradiation per ISO 11137-2
- Non-pyrogenic, non-cytotoxic
- DNase/Rnase, BSE/TSE and human DNA free
Certifications: Compliant with California Prop 65