VWR®, Environmental 90 mm Petri plates
Supplier: VWR Chemicals
Certificates
About this item
Environmental Petri plates 90 mm for active and passive air monitoring in pharmaceutical industries.
- Lateral labelling QR barcode for easy and automatic identification
- Available with broad spectrum disinfectant neutralisers
- H₂O₂ impermeable film for the use inside isolators
- Single, double- or triple-sterile film wrapping
- Gamma irradiated
- Desiccant inside to avoid condensation
- Suitable for use with microbiological air samplers
Long shelf life: Up to 9 months
Room temperature storage: 15 to 25 °C
Environmental 90 mm Petri plates: Three reasons why are they different from standard 90 mm plates.
1) Use: Control of the contamination of air inside cleanrooms, sterile areas and isolators — only for environmental control.
2) Special packaging: Double-/triple-irradiated wrapping is necessary to ensure that the package itself doesn’t contaminate clean/sterile rooms — the first external wrapper is removed just before entering the clean area.
3) For the use inside isolators, a H₂O₂ (VHP) impermeable film is mandatory.
Ordering information: Data logger temperature on delivery available on request.
Related Information
Certificates
Search by Lot NumberSDS_EN_UK_VWRCP10641ZA.pdf SDS_EN_UK_VWRCP11203ZA.pdf SDS_EN_UK_VWRCP11241ZF.pdf SDS_EN_UK_VWRCP21114.pdf SDS_EN_UK_VWRCP30884.pdf
SDS
More
TDS Sabouraud TLHTh AGAR TDS Sabouraud Chloramphenicol Agar TDS Sabouraud Dextrose Agar TDS Sabouraud Dextrose Agar_01 TDS TSA (Tryptone Soya Agar)
Datasheet
More
VWR® HSG Microbiology VWR® HSG Ovens and Incubators