Fetal bovine serum (United States), Hyclone™, standard
Catalog # SH30088.03IH25
Supplier: HyClone products (Cytiva)
Specifications
- Serum format:Frozen
- Sterility:Sterile filtered
- Description:HyClone FBS Standard (US), heat inactivated, irradiated, 500 ml
- Storage temperature:Less than –10 °C
- Pk:500 ml
Specifications
About this item
HyClone standard fetal bovine serum (FBS), U.S. origin is an economical alternative to characterised and defined FBS. Standard FBS is ideal for research applications.
- Sourced in the USA with complete traceability back to original source
- Low in antibodies and high in growth factors
- Produced using true pool technology for uniformity and consistency from bottle to bottle
- Filtered through three sequential 100 nm (0.1 µm) pore size-rated filters
- Virus panel testing according to 9 CFR 113.53
Standard FBS is assayed for gamma globulin, alkaline phosphatase, lactate dehydrogenase, glutamic pyruvic transaminase, glutamic oxaloacetic transaminase, pH, total protein, albumin, blood urea nitrogen, creatinine, total bilirubin, sodium, potassium, calcium, chloride, inorganic phosphorus, osmolality, iron, total iron binding capacity (TIBC) and IgG. Included assays are subject to change without notice.