

Strontium nitrate ≥99.0%, AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:10042-76-9
:
Formula:Sr(NO₃)₂
MDL Number:MFCD00011248
:211.63 g/mol
Merck Index:13,08926
Boiling Pt:645 °C (1013 hPa)
Melting Pt:570 °C
Density:2.113 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:233-131-9
UN:1507
ADR:5.1,III
:GHS03
:GHS05
Specification Test Results
- Assay:min. 99.0%
- Appearance:white crystals
- pH (25°C, 5%):5,5 - 8,0
- Insoluble matter:max. 0.02%
- Loss on drying:max. 0.1%
- Cl (Chloride):max. 20 ppm
- NH4 (Ammonium):max. 100 ppm
- P (Phosphorus):max. 10 ppm
- S (Sulfur):max. 0.05%
- Ag (Silver):max. 5 ppm
- Al (Aluminium):max. 5 ppm
- Ba (Barium):max. 0.05%
- Bi (Bismuth):max. 5 ppm
- Ca (Calcium):max. 70 ppm
- Cd (Cadmium):max. 5 ppm
- Cr (Chromium):max. 5 ppm
- Cu (Copper):max. 5 ppm
- Fe (Iron):max. 30 ppm
- Hg (Mercury):max. 5 ppm
- K (Potassium):max. 100 ppm
- Mg (Magnesium):max. 30 ppm
- Na (Sodium):max. 30 ppm
- Ni (Nickel):max. 5 ppm
- Pb (Lead):max. 5 ppm
- Sb (Antimony):max. 5 ppm
- Si (Silicon):max. 100 ppm
- Sn (Tin):max. 5 ppm
- Ti (Titanium):max. 5 ppm
- Tl (Thalium):max. 5 ppm
- Zn (Zinc):max. 5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC85978.230 spec_VWRC85978.230 spec_VWRC85978.260