Drums, stainless steel
Supplier: Sampling Systems
Certificates
About this item
These stainless steel containers are made from high-quality 316L stainless steel and are ideal for almost all aplications, including GMP use.
- Suitable for pharmaceutical, chemical, food, and cosmetic manufacturing
- Complete with lid, clamp, and FDA compliant seal
- Extremely easy and secure closing and reopening
- High-quality mirror finish
- Fast delivery
This range of stainless steel containers is perfect for dispensing liquids. Due to their ease of use, they are very popular across many different industries.
Ordering information: Emptying tap has to be ordered separately.