Deep-freeze color dots
Supplier: GA
Certificates
About this item
Deep-freeze, 0,28" colours dot labels are designed specifically for the identification of 0,2 ml microtube caps.
- Available in 15 colours, and packs of 15 assorted colours for colours-coding and easy identification
- Labels withstand a wide temperature range (–80 to +110 °C)
- Perfect for storage in laboratory refrigerators and freezers
Waterproof, and with a strong permanent adhesive, these ultra-small diameter deep-freeze labels will resist prolonged freezer storage (–20 °C/–4 °F; –40 °C/–40 °F; –80 °C/–112 °F), refrigeration, and dry ice transport. Lab-TAG™ freezer grade circle labels are ideal for labelling polypropylene tubes and vials as well as other containers, and they can be written on with permanent markers. FDA 21 CFR 175.105 compliant for indirect food contact, they can be used by the food industry for identifying food containers and freezer storage.