Order Entry
Finland
ContactUsLinkComponent
Sink S, b.safe
Sink S, b.safe
Catalog # BOHLR1250-20
Supplier:  Bohlender
undefined
Sink S, b.safe
Catalog # BOHLR1250-20
Supplier:  Bohlender
Specifications
  • Capacity:
    10 L
  • Description:
    b.safe Sink S
  • Connection:
    GL 25 male (bottom)
  • Pk:
    1
  • W×D×H int.:
    333×213×160 mm
Specifications
About this item
Made of static dissipative PE-EX, with sloped bottom for complete emptying.
  • With mounted grounding cable
  • Can be covered completely
To be integrated in laboratory hoods or working tables for an easy disposal of large quantities of liquid solvent wastes. Thread connection GL 25 to connect b.safe waste tubes that transport liquid solvent wastes directly in canisters.
Drain Ø 60 mm with removable sieve. Bottom drain with male thread GL 25 for direct connection to flexible b.safe waste tubes. Grounding connection at the side with mounted grounding cable (1,5 m length).
Accessories information: When not in use, the sink can be covered completely with the separately available Lid (BOHLR1255-20) and used as additional working space.