When grown on CELLvo™ Matrix, CELLvo™ Human Chondrocytes retain their articular cartilage phenotype through more population doublings.
- High col2/col1 ratio
- Elaborate a collagen and GAG rich matrix
- Proliferate rapidly on CELLvo™ Matrix
Human Articular Chondrocytes are mature cells responsible for elaborating and maintaining cartilage matrices in vivo. These cells can be difficult to isolate and expand but have huge potential in translational research. CELLvo™ Chondrocytes are isolated from cadaveric articular cartilage of otherwise healthy donors. CELLvo™ Chondrocytes are intended for research use only and are the ideal choice for researchers.
This product can be used for basic and translation research in the following applications: osteoarthritis research, joint research, chondrocyte differentiation, tissue engineering and chondrocyte implantation studies.
- Storage conditions:Upon arrival, store cells in liquid nitrogen vapour
- Storage buffer:CryoStor®CS5