Order Entry
Finland
ContactUsLinkComponent
Lactic acid 88%, AR®, Macron Fine Chemicals™
Lactic acid 88%, AR®, Macron Fine Chemicals™
Catalog # 3130-46
CAS Number:  
Lactic acid 88%, AR®, Macron Fine Chemicals™
Catalog # 3130-46
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Appearance colorless to light yellow, syrupy liquid:
    Passes test
  • Assay :
    88 - 92 %
  • Reducing sugars passes test passes test:
    Passes test
  • Residue after Ignition (as SO4):
    ≤ 0,04 %
  • Readily Carbonizable Substances:
    Passes test
  • Chloride (Cl):
    ≤ 0,002 %
  • Sulfate (SO4):
    ≤ 0,005 %
  • Heavy Metals (as Pb):
    ≤ 0,0006 %
  • Arsenic (As):
    ≤ 0,0001 %
  • Iron (Fe):
    ≤ 0,0005 %
  • Stereochemical purity (L isomer assay):
    ≥95%

Specification Test Results

About this item

 :
ADR:8,II
 :90.08 g/mol
UN:3265
 :GHS05
CAS Number:50-21-5
 :598-82-3
Formula:H₃CCH(OH)COOH
MDL Number:MFCD00004520