Order Entry
Finland
ContactUsLinkComponent
Kanamycin sulphate 5000 µg/ml
Kanamycin sulphate 5000 µg/ml
Catalog # MDTC30-006-CF
Supplier:  Corning
CAS Number:  
undefined
Kanamycin sulphate 5000 µg/ml
Catalog # MDTC30-006-CF
Supplier:  Corning
CAS Number:  

Specifications

  • Pk:
    6
  • Seq:
    2378
  • Pk Info:
    6×50 ml

Specifications

About this item

 :
 :582.6 g/mol
Storage Temperature:Freezer
CAS Number:25389-94-0
Formula:C₁₈H₃₈N₄O₁₅S
MDL Number:MFCD00070253