Toilet cleaners, SURE®
Supplier: Diversey
Certificates
About this item
Ready to use, heavy-duty cleaner for effective removal of lime scale and other deposits from toilet bowls and urinals.
- Quickly dissolves calcium/lime deposits
- Long lasting action on vertical/sloped surfaces
- Easily reaches hidden areas such as under the toilet rim
- Cleans without leaving residue
- No fragrance or dye
- Contains plant-based ingredients from renewable sources
- Decomposed by natural processes
- No environmental hazard classifications, no safety warnings for product in use for user
SURE® toilet cleaner is part of a unique range of plant-based, cleaning products that deliver the professional cleaning results you need - while being safe for people and kind to the environment. SURE® natural cleaning products provide safety and sustainability without compromising on performance.
Certifications: OECD 301B test.
EU-Ecolabel (EU: NL/020/045).
Detergent Regulation, EC 648/2004.
EU-Ecolabel (EU: NL/020/045).
Detergent Regulation, EC 648/2004.
Specifications
- Catalog No:
- 115-3073
- 115-3072
- Environmentally Preferable:
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging
- Sustainable Materials, Safer Human and Environmental Health, Sustainable Packaging