Order Entry
Finland
ContactUsLinkComponent
TCM defined serum-free growth medium
TCM defined serum-free growth medium
Certificates
undefined
TCM defined serum-free growth medium

Specifications

  • Species:
    Bovine
  • Total protein:
    664 µg/ml at 1X
  • Shelf life:
    18 months from DOM
  • Endotoxin:
    <0,5 EU/ml at 1X
  • Storage temperature:
    2...8 °C

Specifications