SPE columns, PFAS, CHROMABOND
Toimittaja: MACHEREY-NAGEL
Sertifikaatit
Tietoja tästä tuotteesta
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.
- PFAS analysis
- Modern polymer phase
- Excellent recovery rates due to the spherical structure of the particles
- High-purity material with high reproducibility
Polypropylene (PP) columns with polyethylene (PE) filter elements.
Tekniset tiedot
- Catalog No:
- MANA730283
- MANA730283.250
- Application:
- Per- and polyfluoroalkyl substances (PFAS) from water, PFAS from sediments (contaminated soils), PFAS from textiles
- Per- and polyfluoroalkyl substances (PFAS) from water, PFAS from sediments (contaminated soils), PFAS from textiles
- pH range:
- 1,0 - 14,0
- 1,0 - 14,0
- Storage:
- RT
- RT