HPLC columns, PFAS, NUCLEODUR®
Supplier: MACHEREY-NAGEL
Certificates
About this item
Hydrophobic reversed phase with distinct polar selectivity for special PFAS analysis. Suitability for PFAS analysis is assured through qualified batch analysis.
- High purity spherical silica gel
- Polar selectivity
- PFAS analysis
- Low bleeding characteristics
This HPLC phase is made of high purity NUCLEODUR HPLC silica gel and is extensively endcapped. Due to a low bleeding characteristics these columns are suitable for high sensitivity LC/MS analysis.
Delivery: Supplied with a test chromatogram, instruction leaflet and batch test certificate.
Specifications
- Catalog No:
- MANA760666.20
- MANA760663.20
- Application:
- PFAS compounds
- PFAS compounds
- Endcapped:
- Yes
- Yes
- Max. pressure:
- 600 bar
- 600 bar
- pH range:
- 1,0 - 9,0
- 1,0 - 9,0
- Surface area:
- 340 m²/g
- 340 m²/g
- Temperature limit:
- 60 °C
40 °C (phosphate buffer) - 60 °C
40 °C (phosphate buffer)
- Storage:
- RT
- RT
- Shipping conditions:
- Eluent in HPLC column
Acetonitrile – water - Eluent in HPLC column
Acetonitrile – water