Order Entry
Finland
ContactUsLinkComponent
General purpose gloves, Perfect Poly® Workeasy Black black
General purpose gloves, Perfect Poly® Workeasy Black black
Supplier:  HONEYWELL SAFETY
Certificates
undefined
General purpose gloves, Perfect Poly® Workeasy Black black
Supplier:  HONEYWELL SAFETY
Specifications
  • Catalog No:
  • 112-1634
  • 112-1633
  • 112-1631
  • 112-1632
  • Model:
  • Workeasy Black Black
  • Workeasy Black Black
  • Workeasy Black Black
  • Workeasy Black Black
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Polyester
  • Polyester
  • Polyester
  • Polyester
  • Finish:
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Palm and fingertips coated
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (2121X)
  • EN 388:2016 (2121X)
  • EN 388:2016 (2121X)
  • EN 388:2016 (2121X)
Specifications