VWR®, Peristaltic Pump, Media
Catalog # 710-2126
Supplier: VWR Collection
Specifications
- Flow rate:1,5 - 1250 ml/min
- Rotation speed:10 - 550 min⁻¹
- Description:Media pump, with single pump head and 3 rollers, for culture preparation
- Model:VWR® media pump, single head
- Electrical:100 - 240 V, 50 - 60 Hz
- Int. Ø tubing:1,6; 3,2; 4,8; 6,4; 8 mm
- Weight:5,4 kg
- W×D×H:161×273×208 mm
- Pk:1
Specifications
About this item
Peristaltic pump for culture media preparation. Use it for serial dispensing of culture media, agar or any liquid diluent for efficient sample preparation.
- Select from three dispensing modes: Continuous, dose (pre-determined volume), multi-dose (pre-determined volume, with an adjustable pause between each dose)
- Stainless steel and heavy-duty polycarbonate top shell
- Rotating button for easy navigation
- Compact and ergonomic: Only 160 mm wide
- Typical precision with inner Ø 3,2 mm tubing: 9 ml: ±1,8%, 18 ml: ±1%, 225 ml: ±0,5%
- Dispenses liquid from 50 μl - 99 L
- Improved dispensing control with the handy gun and the adjustable arm
- User-friendly interface
- High quality Watson Marlow™ pump heads
Certifications: In compliance with ISO 7218 and FDA BAM (Bacteriological Analytical Manual).
Delivery: Pump is delivered with peristaltic Watson Marlow™ pump head, 1 set of 3,2 mm dispensing assembly (nozzle + GL 45 bottle connector + filter + tubing + tube weight), 1 set of power cables (EU, UK, CH), user’s manual, and conformity certificate.