Brilliant II probe MM
Supplier: AGILENT
Certificates
About this item
Brilliant II QPCR and QRT-PCR reagents were developed for improved sensitivity of detection, ensuring reproducible quantification even at low target concentrations.
- Greater flexibility for use with numerous different templates and targets
- Developed on the Mx QPCR Systems
- Passive reference dye to maximise performance on different instrument platforms
- Multiplex kit for improved results on multiple targets
For faster experiments with earlier Ct's and greater reproducibility, Brilliant III Ultra-Fast master mixes can be used.
Caution: For research use only. Not for use in diagnostic procedures.
Specifications
- Application:RT-qPCR probe
- One step:Yes