Order Entry
Finland
ContactUsLinkComponent
Spin concentrators, 50K MWCo
Spin concentrators, 50K MWCo
Catalog # AGLS5188-5202
Supplier:  AGILENT
Spin concentrators, 50K MWCo
Catalog # AGLS5188-5202
Supplier:  AGILENT

Specifications

  • MWCO:
    50 K
  • Description:
    Spin concentrators, 4 ml
  • Pk:
    25

Specifications

About this item

Spin concentrators are devices used to concentrate protein samples via low-speed centrifugation through MWCo filters into a collection tube.

  • Contains a polyethersulfone (PES) membrane that is compatible with biological fluids and most aqueous solutions with pore sizes of 50 kDa
  • Active membrane area of 2,0 or 4,0 cm²
  • Durable and designed to minimise membrane fouling and allow fast concentration, even with particles in the solution

Designed to fit a standard 15 ml conical collection tube or a 50 ml collection tube (not provided with concentrator). Designed to be used in either swing-bucket or fixed angle rotor centrifuge equipment at maximum 5000 or 2000 g.