Order Entry
Finland
ContactUsLinkComponent
Resin, StrataClean
Resin, StrataClean
Supplier:  AGILENT
Certificates
Resin, StrataClean
Supplier:  AGILENT

Specifications

  • Method / Format:
    Affinity resin
  • Prep size:
    Scalable
  • Refine target molecule:
    DNA preps
    Total DNA
    Amplified/modified DNA
  • Sample type:
    DNA preps
    PCR and other enzymatic reactions
  • Target molecule:
    DNA

Specifications