Multiple affinity removal column HSA/IgG
Affinity removal system column, human albumin/IgG, 4,60 mm, 50 mm
AGLS5188-8826
: AGILENT
The multiple affinity removal LC column – human albumin/IgG is designed to chromatographically remove the two most interfering high-abundant proteins (albumin and IgG) from human samples (such as plasma, serum, urine or cerebrospinal fluid).
- Serum injection volume is 100 µl per run for the 4,6 mmid column
- Optimise sample recovery by utilising rapid, single injection protocols and optimised buffers
- Column is compatible with most FPLC or HPLC pumps or instruments
Removal of albumin and IgG improves the subsequent LC/MS and electrophoretic analysis of the sample by effectively expanding the dynamic range of the analysis.
- Ø int.:4,60 mm
- Description:Affinity removal system column, human albumin/IgG
- Length:50 mm



