Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant HIV-1 gp120 CM
Recombinant HIV-1 gp120 CM
APOSBITP1885-10UG
:
Apollo Scientific
:
Recombinant HIV-1 gp120 CM
APOSBITP1885-10UG
:
Apollo Scientific
:
Pk:
10 µG
Protein/peptide type:
Recombinant
Seq:
05
Protein/peptide name:
HIV-1 gp120 CM