Order Entry
Finland
ContactUsLinkComponent
Recombinant Hepatitis B Surface Antigen adr subtype
Recombinant Hepatitis B Surface Antigen adr subtype
Catalog # APOSBITP1834-50UG
Supplier:  Apollo Scientific
CAS Number:  
Recombinant Hepatitis B Surface Antigen adr subtype
Catalog # APOSBITP1834-50UG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Protein synonyms:
    HBsAg adr
  • Protein/peptide name:
    Hepatitis B Surface Antigen adr subtype

Specifications

About this item