Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Hepatitis C Virus NS3 Genotype-5a
Recombinant Hepatitis C Virus NS3 Genotype-5a
APOSBITP1771-1MG
:
Apollo Scientific
:
Recombinant Hepatitis C Virus NS3 Genotype-5a
APOSBITP1771-1MG
:
Apollo Scientific
:
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
HCV NS3 Genotype-5a
Protein/peptide name:
Hepatitis C Virus NS3 Genotype-5a