Order Entry
Finland
ContactUsLinkComponent
Bovine Holo Transferrin
Bovine Holo Transferrin
Catalog # APOSBITP1578-1G
Supplier:  Apollo Scientific
CAS Number:  
Bovine Holo Transferrin
Catalog # APOSBITP1578-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pk:
    1 g
  • Species:
    Bovine
  • Protein/peptide name:
    Holo Transferrin

Specifications

About this item