Order Entry
Finland
ContactUsLinkComponent
 
Human Recombinant Leptin (from E. coli)
 
Human Recombinant Leptin (from E. coli)

 

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Protein/peptide name:
    Leptin

 

Frequently Bought Together