Mouse recombinant KC
Catalog # APOSBITC2011-1MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pk:1 mg
- Protein/peptide type:Recombinant
- Source:
- Species:Mouse
- Environmentally Preferable:
- Protein synonyms:GRO/KC, CXCL1
- Protein/peptide name:KC