NANOCOLOR® Photometric Tube Tests, Phenol and Accessories
Supplier: MACHEREY-NAGEL
Certificates
About this item
Photometric test kit for the determination of phenol in water samples and accessories for the round cell test NANOCOLOR® Phenol.
- Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
- Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
- Precisely pre-dosed reagents with constant high quality from batch to batch
- Colored pictogram instructions and easy handling of the tubes
- 16 mm (OD) tubes for easy pipetting
The test kit is evaluated in 16 mm (AD) tubes. It is pre-programmed on NANOCOLOR® photometers and is also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.
Hazardous material: Yes
Sludge reagent set: No (except: MANA918929)
Digestion set: No (except: MANA918929)
NanOx Metal: No (except: MANA918929)
NanOx N: No (except: MANA918929)
Specifications
- Model:MANA985074
- Platform:NANOCOLOR® tube tests
- Storage temperature:15...25 °C
- Shelf life (months):18
- Weight:348 g (Including packaging)
Specifications
Related Information
Frequently Bought Together