Reducing tubing connectors, b.safe
Supplier: Bohlender
Certificates
About this item
Straight fitting made of PP, with two hose connectors of elastic tubing with different inner diameters.
- Excellent chemical resistance
- Available for a wide range of hose diameters
- The conical shape gets the connection tight and leak-free
b.safe hose connectors are the flexible choice to customize your b.safe waste cap. Made of PP or PTFE and available in numerous diameters you will find the exact one to fit your waste hose.