About this item
Filter body made of PP with connector (thread GL 14) for b.safe waste caps. Lid with aeration openings and service life indicator with additional label field to monitor service life. Filling made of activated carbon for adsorption of solvents.
- Activation of service life through touch of the button and/or individual marking on the back side
- Splash-proof ventilation openings
- Activated carbon in the inside guarantee adsorption of harmful vapours
- PP for high resistance and fire class rating V-0
- Plenty of space for fittings and hose connectors
Safe collection of solvents without leakage of harmful solvent vapours. The activated carbon in the b.safe exhaust filter restrains solvent vapours from the escaping air during filling. The service life of the filter can be easily monitored through either activating the display on the lid and exchanging the filter in case of completely red colouring or documentation of the exchange date by hand. We recommend the following filter sizes: Size small for up to 5 L, size medium for 5 to 10 L, and size large for more than 10 L.