Order Entry
Finland
ContactUsLinkComponent
VWR®, Cut Resistant Gloves, NBR Coating
VWR®, Cut Resistant Gloves, NBR Coating
Supplier:  VWR Collection
Certificates
undefined
VWR®, Cut Resistant Gloves, NBR Coating
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • 112-5682
  • 112-5681
  • 112-5684
  • 112-5683
  • 112-5680
  • 112-5679
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Reusable
  • Latex / latex-free:
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Latex-free
  • Finish:
  • Nitrile (NBR) palm coating
  • Nitrile (NBR) palm coating
  • Nitrile (NBR) palm coating
  • Nitrile (NBR) palm coating
  • Nitrile (NBR) palm coating
  • Nitrile (NBR) palm coating
  • Gauge:
  • 13
  • 13
  • 13
  • 13
  • 13
  • 13
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)
  • EN 388:2016 (4X42B)