Order Entry
Finland
ContactUsLinkComponent
Tubing holder, b.safe
Tubing holder, b.safe
Catalog # 590-0220
Supplier:  Bohlender
undefined
Tubing holder, b.safe
Catalog # 590-0220
Supplier:  Bohlender

Specifications

  • Description:
    Tubing holder for 8× Ext.Ø 1,6 mm; 3× Ext.Ø 3,2 mm or 1× Ext.Ø 5 mm
  • Length:
    30 mm
  • Pk:
    10

Specifications

About this item

For fixation of capillaries and tubing to avoid disarranging or bending of tubes. Self-adhesive bottom side.

  • Avoid disarranging or bending of tubes
  • Lead and fix b.safe tubes
  • Made of ABS

A b.safe tubing holder can arrange at once up to 8× Ext.Ø 1,6 mm / 3× Ext.Ø 3,2 mm or 1× Ext.Ø 5 mm.

To lead and fix tubing and capillaries without being disarranged or bent.