Politainers, b.safe
Supplier: Bohlender
Certificates
About this item
Foldable canister for the collection of liquid waste.
- Including screw cap
- Foldable canister
Folded to save space during delivery and storage, unfolds automatically during filling. UN-approval only if used with the appropriate carton box.
Ordering information: Please order additional accessories like the carton box or the adaptor for Waste Caps GL 45 separately.