Methyl-4-chloro-6-oxo-1-[3-(trifluoromethyl)phenyl]-1,6-dihydro-3-pyridazinecarboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:129109-18-8
:
Formula:C₁₃H₈ClF₃N₂O₃
MDL Number:MFCD00141148
:332.67 g/mol
Melting Pt:98…100 °C
Storage Temperature:Ambient