Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(Trifluoromethyl)phenylhydrazinium chloride
2-(Trifluoromethyl)phenylhydrazinium chloride
2-(Trifluoromethyl)phenylhydrazinium chloride
:
Apollo Scientific
2-(Trifluoromethyl)phenylhydrazinium chloride
:
Apollo Scientific
:
3107-34-4
:
:
C₇H₈ClF₃N₂
:
MFCD00102619
:
212.6
:
220 °C
:
GHS07
:
Ambient