2-Bromo-4,5-difluoroaniline 97%
Catalog # APOSPC900805-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:208.0000 g/mol
Storage Temperature:Ambient
CAS Number:64695-79-0
Formula:C₆H₄BrF₂N
MDL Number:MFCD04112491