Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Benzyloxy-1-bromo-2-fluorobenzene
4-Benzyloxy-1-bromo-2-fluorobenzene 98%
4-Benzyloxy-1-bromo-2-fluorobenzene 98%
:
Apollo Scientific
4-Benzyloxy-1-bromo-2-fluorobenzene 98%
:
Apollo Scientific
:
185346-79-6
:
:
C₁₃H₁₀BrFO
:
MFCD09801041
:
281.13 g/mol
:
33…35 °C
:
GHS07
:
Ambient